missing translation for 'onlineSavingsMsg'
Learn More

cis,cis-1,5-Cyclooctadiene, 99%, stab. with 50-200ppm Irganox 1076

Catalog No. AAB21196AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21196AU 1000 mL
AAB21196AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB21196AU Supplier Thermo Scientific Chemicals Supplier No. B21196AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1552-12-1 | C8H12 | 108.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1552-12-1
Molecular Formula C8H12
Molecular Weight (g/mol) 108.184
MDL Number MFCD00001752
InChI Key VYXHVRARDIDEHS-QGTKBVGQSA-N
Synonym 1,5-cyclooctadiene, cycloocta-1,5-diene, 1z,5z-cycloocta-1,5-diene, 1,5-cyclooctadiene z,z, 1z,5z-cyclooctadiene, z,z-cycloocta-1,5-diene, 1-cis,5-cis-cyclooctadiene, unii-1e1vvd385z, cis,cis-cycloocta-1,5-diene, 1,5-cod
PubChem CID 82916
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene
SMILES C1CC=CCCC=C1

Specifications

Melting Point -70°C
Density 0.88
Boiling Point 151°C to 152°C
Flash Point 32°C (89°F)
Odor Unpleasant
Assay Percent Range 0.99
Refractive Index 1.494
Quantity 1000 mL
UN Number UN2520
Beilstein 1209288
Sensitivity Air Sensitive
Formula Weight 108.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material cis,cis-1,5-Cyclooctadiene, stabilized with 50 to 200ppm Irganox 1076
Show More Show Less
Hazard Category H226-H302+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H226-H302-H332-H315-H319-H317-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CYCLOOCTADIENES
EINECSNumber 216-291-4
RTECSNumber GX9620000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only