missing translation for 'onlineSavingsMsg'
Learn More

cis,cis-1,3,5-Cyclohexanetriol dihydrate, 98%, Thermo Scientific Chemicals

Catalog No. ac458950050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458950050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458950050 Supplier Thermo Scientific Chemicals Supplier No. 458950050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60662-54-6
Molecular Formula C6H12O3·2H2O
Molecular Weight (g/mol) 168.19
InChI Key MAPLMYMZLRIJSP-UHFFFAOYSA-N
Synonym cis,cis-1,3,5-cyclohexanetriol dihydrate, cis,cis-1,3,5-trihydroxycyclohexane, cis-phloroglucitol, 1s,3s,5s-cyclohexane-1,3,5-triol dihydrate, ciscis-135-cyclohexanetrioldihydrate, cyclohexane-1,3,5-triol dihydrate, cyclohexane-1,3,5-triol-water 1/2, cis cis-1 3 5-cyclohexanetriol dihydrate, 1r,3s,5s-cyclohexane-1,3,5-triol dihydrate
PubChem CID 16211857
IUPAC Name cyclohexane-1,3,5-triol;dihydrate
SMILES C1C(CC(CC1O)O)O.O.O

Specifications

Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Solubility Information Solubility in water: soluble in hot water.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material cis,cis-1,3,5-Cyclohexanetriol dihydrate

RUO – Research Use Only