missing translation for 'onlineSavingsMsg'
Learn More

cis,cis-1,3,5-Cyclohexanetriol dihydrate, 98%, Thermo Scientific™

Catalog No. ac458950010
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458950010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC458950010 Supplier Thermo Scientific Chemicals Supplier No. 458950010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 60662-54-6
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H12O3·2H2O
Quantity 1g
Synonym cis,cis-1,3,5-cyclohexanetriol dihydrate, cis,cis-1,3,5-trihydroxycyclohexane, cis-phloroglucitol, 1s,3s,5s-cyclohexane-1,3,5-triol dihydrate, ciscis-135-cyclohexanetrioldihydrate, cyclohexane-1,3,5-triol dihydrate, cyclohexane-1,3,5-triol-water 1/2, cis cis-1 3 5-cyclohexanetriol dihydrate, 1r,3s,5s-cyclohexane-1,3,5-triol dihydrate
Solubility Information Solubility in water: soluble in hot water.
InChI Key MAPLMYMZLRIJSP-UHFFFAOYSA-N
SMILES C1C(CC(CC1O)O)O.O.O
IUPAC Name cyclohexane-1,3,5-triol;dihydrate
Molecular Weight (g/mol) 168.19
PubChem CID 16211857
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material cis,cis-1,3,5-Cyclohexanetriol dihydrate
Show More Show Less

RUO – Research Use Only