missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) dihydrate, Ru 19% min
Description
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) dihydrate is used in the preparation of [Ru(m-) 2,2'-bipyridine)3]2+(PF6-)2. It is also involved in the preparation of an intermediate for the development of nanocomposite junctions with gold nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| Assay Percent Range | Ru 19% min. |
| Quantity | 1 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 520.38 (484.35 Anhydrous) |
| Physical Form | Crystalline |
| Chemical Name or Material | cis-Bis-(2, 2 ft.-bipyridine)dichlororuthenium(II) dihydrate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only