missing translation for 'onlineSavingsMsg'
Learn More

cis-9-Tricosene 98.0+%, TCI America™

$87.60 - $305.70

Chemical Identifiers

CAS 27519-02-4
Molecular Formula C23H46
Molecular Weight (g/mol) 322.621
MDL Number MFCD00008988
InChI Key IGOWHGRNPLFNDJ-ZPHPHTNESA-N
Synonym cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
PubChem CID 5365075
IUPAC Name (Z)-tricos-9-ene
SMILES CCCCCCCCCCCCCC=CCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T12421ML
View Documents
TCI America
T12421ML
1 mL N/A
Only null left
 
T12425ML
View Documents
TCI America
T12425ML
5 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

27519-02-4
322.621
IGOWHGRNPLFNDJ-ZPHPHTNESA-N
5365075
CCCCCCCCCCCCCC=CCCCCCCCC
C23H46
MFCD00008988
cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
(Z)-tricos-9-ene

Specifications

27519-02-4
C23H46
1 mL
IGOWHGRNPLFNDJ-ZPHPHTNESA-N
(Z)-tricos-9-ene
5365075
≥98.0% (GC)
cis-9-Tricosene
Yellow
MFCD00008988
cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
CCCCCCCCCCCCCC=CCCCCCCCC
322.621
322.62
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : YD0807000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'