missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-9-Tricosene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 mL |
| Formula Weight | 322.62 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | cis-9-Tricosene |
Safety and Handling
| RTECSNumber | YD0807000 |
| TSCA | Yes |