missing translation for 'onlineSavingsMsg'
Learn More

cis-9-Tricosene 98.0+%, TCI America™

Catalog No. T12421ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12421ML 1 mL
T12425ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12421ML Supplier TCI America Supplier No. T12421ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27519-02-4
Molecular Formula C23H46
Molecular Weight (g/mol) 322.621
MDL Number MFCD00008988
InChI Key IGOWHGRNPLFNDJ-ZPHPHTNESA-N
Synonym cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
PubChem CID 5365075
IUPAC Name (Z)-tricos-9-ene
SMILES CCCCCCCCCCCCCC=CCCCCCCCC

Specifications

Color Yellow
Quantity 1 mL
Formula Weight 322.62
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-9-Tricosene
RTECSNumber YD0807000
TSCA Yes