missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-9-Hexadecenoic Acid 98.0+%, TCI America™
$108.21 - $472.75
Chemical Identifiers
| CAS | 373-49-9 |
|---|---|
| Molecular Formula | C16H30O2 |
| Molecular Weight (g/mol) | 254.41 |
| MDL Number | MFCD00004437 |
| InChI Key | SECPZKHBENQXJG-FPLPWBNLSA-N |
| Synonym | palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid |
| PubChem CID | 445638 |
| ChEBI | CHEBI:28716 |
| IUPAC Name | (9Z)-hexadec-9-enoic acid |
| SMILES | CCCCCC\C=C/CCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H0072100MG
|
TCI America
H0072100MG |
100 mg |
Each for $108.21
|
|
|||||
|
H00721G
|
TCI America
H00721G |
1 g |
Each for $472.75
|
|
|||||
Chemical Identifiers
| 373-49-9 | |
| 254.41 | |
| SECPZKHBENQXJG-FPLPWBNLSA-N | |
| 445638 | |
| (9Z)-hexadec-9-enoic acid |
| C16H30O2 | |
| MFCD00004437 | |
| palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid | |
| CHEBI:28716 | |
| CCCCCC\C=C/CCCCCCCC(O)=O |
Specifications
| 373-49-9 | |
| Yellow | |
| C16H30O2 | |
| 100 mg | |
| SECPZKHBENQXJG-FPLPWBNLSA-N | |
| (9Z)-hexadec-9-enoic acid | |
| 445638 | |
| 254.41 | |
| Liquid |
| 1°C | |
| 175°C | |
| MFCD00004437 | |
| palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid | |
| CCCCCC\C=C/CCCCCCCC(O)=O | |
| 254.41 | |
| CHEBI:28716 | |
| ≥98.0% (GC,T) | |
| cis-9-Hexadecenoic Acid |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : No
Recommended Storage : Refrigerator