missing translation for 'onlineSavingsMsg'
Learn More

cis-9-Hexadecenoic Acid 98.0+%, TCI America™

Catalog No. H0072100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0072100MG 100 mg
H00721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0072100MG Supplier TCI America Supplier No. H0072100MG

Chemical Identifiers

CAS 373-49-9
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00004437
InChI Key SECPZKHBENQXJG-FPLPWBNLSA-N
Synonym palmitoleic acid, cis-9-hexadecenoic acid, z-hexadec-9-enoic acid, 9-cis-hexadecenoic acid, zoomaric acid, z-9-hexadecenoic acid, 9z-hexadecenoic acid, palmitolinoleic acid, cis-palmitoleic acid, zoomeric acid
PubChem CID 445638
ChEBI CHEBI:28716
IUPAC Name (9Z)-hexadec-9-enoic acid
SMILES CCCCCC\C=C/CCCCCCCC(O)=O

Specifications

Melting Point 1°C
Color Yellow
Boiling Point 175°C
Quantity 100 mg
Formula Weight 254.41
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material cis-9-Hexadecenoic Acid
EINECSNumber (2)-0609
TSCA No
Recommended Storage Refrigerator