missing translation for 'onlineSavingsMsg'
Learn More

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 97%

Catalog No. p-7027893
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2495106 5 g
AAB2495114 25 g
AAB2495122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2495106 Supplier Thermo Scientific Chemicals Supplier No. B2495106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 129-64-6 | C9H8O3 | 164.16 g/mol

cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

Solubility
It decomposes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 129-64-6
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106
InChI Key KNDQHSIWLOJIGP-UHFFFAOYNA-N
Synonym cis-5-norbornene-endo-2,3-dicarboxylic anhydride
PubChem CID 126963424
IUPAC Name 4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
SMILES O=C1OC(=O)C2C3CC(C=C3)C12

Specifications

Melting Point 164°C to 167°C
Density 1.417
Quantity 5 g
Beilstein 83657
Sensitivity Moisture sensitive
Merck Index 14,1796
Solubility Information It decomposes in water.
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic anhydride
Hazard Category H317-H318-H334
Hazard Statement GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c
EINECSNumber 204-957-7
RTECSNumber DT5600000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only