missing translation for 'onlineSavingsMsg'
Learn More

cis-4-tert-Butylcyclohexanol 98.0+%, TCI America™

Catalog No. B12701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12701G 1 g
B12705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B12701G Supplier TCI America Supplier No. B12701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 937-05-3
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00070476
InChI Key CCOQPGVQAWPUPE-UHFFFAOYSA-N
PubChem CID 7391
IUPAC Name 4-tert-butylcyclohexan-1-ol
SMILES CC(C)(C)C1CCC(CC1)O

Specifications

Melting Point 83°C
Color White-Yellow
Quantity 1 g
Formula Weight 156.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-tert-Butylcyclohexanol