missing translation for 'onlineSavingsMsg'
Learn More

cis-4-tert-Butylcyclohexanecarboxylic Acid 98.0+%, TCI America™

Catalog No. B26315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26315G 5 g
B263125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B26315G Supplier TCI America Supplier No. B26315G
Only null left

Chemical Identifiers

CAS 943-28-2
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
InChI Key QVQKEGYITJBHRQ-UHFFFAOYSA-N
PubChem CID 136759
IUPAC Name 4-tert-butylcyclohexane-1-carboxylic acid
SMILES CC(C)(C)C1CCC(CC1)C(=O)O

Specifications

Melting Point 117°C
Color White
Quantity 5 g
Formula Weight 184.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-tert-Butylcyclohexanecarboxylic Acid