missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Methylcyclohexanol 98.0+%, TCI America™

$77.00 - $232.95

Chemical Identifiers

CAS 7731-28-4
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00064170
InChI Key MQWCXKGKQLNYQG-UHFFFAOYSA-N
Synonym 4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
PubChem CID 11524
IUPAC Name 4-methylcyclohexan-1-ol
SMILES CC1CCC(CC1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M03531ML
View Documents
TCI America
M03531ML
1 mL
Each for $77.00
Only null left
 
M03535ML
View Documents
TCI America
M03535ML
5 mL
Each for $232.95
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7731-28-4
114.188
MQWCXKGKQLNYQG-UHFFFAOYSA-N
11524
CC1CCC(CC1)O
C7H14O
MFCD00064170
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
4-methylcyclohexan-1-ol

Specifications

7731-28-4
171°C
MFCD00064170
4-methylcyclohexanol, trans-4-methylcyclohexanol, cis-4-methylcyclohexanol, cyclohexanol, 4-methyl, 1-methyl-4-cyclohexanol, cyclohexanol, 4-methyl-, trans, methyl cyclohexanol, p-methylcyclohexanol, cyclohexanol, 4-methyl-, cis, trans 4-methyl cyclohexanol
CC1CCC(CC1)O
114.188
114.19
Liquid
Colorless
C7H14O
1 mL
MQWCXKGKQLNYQG-UHFFFAOYSA-N
4-methylcyclohexan-1-ol
11524
≥98.0% (GC)
cis-4-Methylcyclohexanol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2301

TSCA : No

SDS
Documents

Documents