missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4-Methyl-2-pentene 93.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 56°C |
| Quantity | 5 mL |
| UN Number | 2288 |
| Formula Weight | 84.16 |
| Percent Purity | ≥93.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | cis-4-Methyl-2-pentene |
Safety and Handling
| EINECSNumber | (2)-0031 |
| TSCA | No |
| Recommended Storage | Refrigerator |