missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-L-proline 97.0+%, TCI America™

Catalog No. p-7130907
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1169100MG 100 mg
H11691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1169100MG Supplier TCI America Supplier No. H1169100MG
Only null left

Chemical Identifiers

CAS 618-27-9
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00064319,MFCD00064320
InChI Key PMMYEEVYMWASQN-IMJSIDKUSA-N
Synonym cis-4-hydroxy-l-proline, 2s,4s-4-hydroxypyrrolidine-2-carboxylic acid, l-hydroxyproline, l-allohydroxyproline, allo-4-hydroxy-l-proline, cis-4-hydroxyproline, 4s-4-hydroxy-l-proline, l-cis-4-hydroxyproline, l-allo-hydroxyproline, l-proline, 4-hydroxy-, 4s
PubChem CID 440015
ChEBI CHEBI:28397
IUPAC Name (2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@@H]1CN[C@@H](C1)C(O)=O

Specifications

Melting Point 241°C
Color White-Yellow
Quantity 100 mg
Formula Weight 131.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-Hydroxy-L-proline
EINECSNumber (9)-1628
TSCA Yes