missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-D-proline 98.0+%, TCI America™

Catalog No. H13581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13581G 1 g
H13585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13581G Supplier TCI America Supplier No. H13581G

Chemical Identifiers

CAS 2584-71-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00005252
InChI Key PMMYEEVYMWASQN-QWWZWVQMSA-N
Synonym cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid
PubChem CID 440014
ChEBI CHEBI:16231
IUPAC Name (2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@H]1CN[C@H](C1)C(O)=O

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 1 g
Formula Weight 131.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-Hydroxy-D-proline