missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals cis-4-Hydroxy-D-proline, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass Bottle |
| Beilstein | 22, I, 546 |
| Specific Rotation | +56° to +60° (24°C, 589nm) (c=2, water) |
| Merck Index | 10, 4762 |
| Specific Rotation Condition | +58° (24°C c=2,water) |
| Quantity | 1 g |
| Formula Weight | 131.13 |
| Show More |
RUO – Research Use Only