missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cis-4-Hydroxy-D-proline, 99%

Catalog No. AC204910100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC204910100 10 g Glass Bottle
AC204910010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC204910100 Supplier Thermo Scientific Chemicals Supplier No. 204910100

CAS: 2584-71-6 | C5H9NO3 | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2584-71-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00005252
InChI Key PMMYEEVYMWASQN-QWWZWVQMSA-N
Synonym cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid
PubChem CID 440014
ChEBI CHEBI:16231
IUPAC Name (2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@H]1CN[C@H](C1)C(O)=O

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Beilstein 22, I, 546
Specific Rotation +56° to +60° (24°C, 589nm) (c=2, water)
Merck Index 10, 4762
Specific Rotation Condition +58° (24°C c=2,water)
Quantity 10 g
Formula Weight 131.13
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material cis-4-Hydroxy-D-proline
Show More Show Less

RUO – Research Use Only