missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Hydroxy-D-proline, 98+%, Thermo Scientific Chemicals

Catalog No. AAH2763303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2763303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2763303 Supplier Thermo Scientific Chemicals Supplier No. H2763303

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2584-71-6
Molecular Formula C5H9NO3
Molecular Weight (g/mol) 131.13
MDL Number MFCD00005252
InChI Key PMMYEEVYMWASQN-QWWZWVQMSA-N
Synonym cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid
PubChem CID 440014
ChEBI CHEBI:16231
IUPAC Name (2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
SMILES O[C@H]1CN[C@H](C1)C(O)=O

Specifications

Melting Point 243°C (decomposition)
Quantity 1 g
Formula Weight 131.13
Percent Purity ≥98%
Chemical Name or Material cis-4-Hydroxy-D-proline
EINECSNumber 219-963-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only