missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Decenal, 95%

Catalog No. aal0638203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0638203 1 g
AAL0638206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0638203 Supplier Thermo Scientific Chemicals Supplier No. L0638203
Only null left

CAS: 21662-09-9 | C10H18O | 154.253 g/mol

cis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-Decenal is suitable for use as standard compound to investigate the release of odorants in hydrocolloid model systems containing original or regio-selectively carboxylated cellulose at different pH values using static headspace gas chromatographic (SHS-GC) analysis. It may be used as standard for the standardization of 9-decenal by gas-chromatography. It may be used in the preparation of cis-4-decenol. It has been identified as one of the volatile biological marker for grey mold (Botrytis cinerea) infections in strawberry.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 21662-09-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00007024
InChI Key CWRKZMLUDFBPAO-VOTSOKGWSA-N
Synonym trans-4-decen-1-al, trans-4-decenal, e-4-decenal, e-dec-4-enal, 4-decenal, e, 4-decenal, unii-kz74b5801j, 4-decene-1-al, dec-4-enal, fema no. 3264
PubChem CID 5702654
IUPAC Name (E)-dec-4-enal
SMILES CCCCCC=CCCC=O

Specifications

Density 0.857
Boiling Point 78°C to 80°C (10 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.444
Quantity 1 g
Beilstein 2323646
Solubility Information Not miscible in water.
Formula Weight 154.25
Percent Purity 95%
Chemical Name or Material cis-4-Decenal
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 244-514-5
RTECSNumber HE2071400
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only