missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Cyclohexene-1,2-dicarboxylic acid, 98%

Catalog No. AAL0939222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0939222 100 g
AAL0939214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0939222 Supplier Thermo Scientific Chemicals Supplier No. L0939222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2305-26-2 | C8H10O4 | 170.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2305-26-2
Molecular Formula C8H10O4
Molecular Weight (g/mol) 170.164
MDL Number MFCD00070482
InChI Key ILUAAIDVFMVTAU-OLQVQODUSA-N
Synonym cis-4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 4-cyclohexene-1,2-dicarboxylic acid, cis, cis-delta4-tetrahydrophthalic acid, 4-cyclohexene-1,2-dicarboxylic acid, 1r,2s-rel, rel-1r,2s-cyclohex-4-ene-1,2-dicarboxylic acid, 1r,2s-rel-4-cyclohexene-1,2-dicarboxylic acid, cis-.delta.4-tetrahydrophthalic acid, cis-.delta.4-cyclohexene-1,2-dicarboxylic acid
PubChem CID 504243
IUPAC Name (1S,2R)-cyclohex-4-ene-1,2-dicarboxylic acid
SMILES C1C=CCC(C1C(=O)O)C(=O)O

Specifications

Melting Point 165°C to 170°C
Quantity 100 g
Beilstein 2050419
Formula Weight 170.17
Percent Purity 98%
Chemical Name or Material cis-4-Cyclohexene-1,2-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-974-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only