missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Boc-amino)cyclohexaneacetaldehyde, 97%, Thermo Scientific Chemicals

$370.71 - $370.71

Chemical Identifiers

CAS 847417-37-2
Molecular Formula C13H23NO3
Molecular Weight (g/mol) 241.331
MDL Number MFCD03844591
InChI Key OPGBSEXLSWYFOR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
PubChem CID 24720884
IUPAC Name tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CC=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6209903
View Documents
Thermo Scientific Chemicals
H6209903
1 g
Each for $370.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

847417-37-2
241.331
OPGBSEXLSWYFOR-UHFFFAOYSA-N
24720884
CC(C)(C)OC(=O)NC1CCC(CC1)CC=O
C13H23NO3
MFCD03844591
tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate

Specifications

847417-37-2
MFCD03844591
OPGBSEXLSWYFOR-UHFFFAOYSA-N
tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
241.331
241.32
cis-4-(Boc-amino)cyclohexaneacetaldehyde
C13H23NO3
tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)CC=O
1 g
24720884
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only