missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Boc-amino)cyclohexaneacetaldehyde, 97%, Thermo Scientific Chemicals

Catalog No. AAH6209903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6209903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6209903 Supplier Thermo Scientific Chemicals Supplier No. H6209903
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 847417-37-2
Molecular Formula C13H23NO3
Molecular Weight (g/mol) 241.331
MDL Number MFCD03844591
InChI Key OPGBSEXLSWYFOR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-2-oxoethyl cyclohexylcarbamate, tert-butyl cis-4-2-oxoethyl cyclohexylcarbamate, tert-butyl n-4-2-oxoethyl cyclohexyl carbamate, carbamic acid,n-trans-4-2-oxoethyl cyclohexyl-, 1,1-dimethylethyl ester, opgbsexlswyfor-xypyzodxsa-n, tert-butyl 4-2-oxoethyl cyclohexyl carbamate, cis-tert-butyl 4-2-oxoethyl cyclohexyl carbamate, trans-tert-butyl-4-2-oxoethyl-cyclohexylcarbamate, cis-4-tert-butoxycarbonylamino cyclohexylacetaldehyde, tert-butyl trans-4-2-oxoethyl cyclohexyl carbamate
PubChem CID 24720884
IUPAC Name tert-butyl N-[4-(2-oxoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CC=O

Specifications

Quantity 1 g
Formula Weight 241.32
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexaneacetaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only