missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid 97.0+%, TCI America™

Catalog No. B35181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35181G 1 g
B35185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35181G Supplier TCI America Supplier No. B35181G

Chemical Identifiers

CAS 222530-33-8
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD03453252
InChI Key JSGHMGKJNZTKGF-BDAKNGLRSA-N
Synonym 1r,3s-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino-cyclohexanecarboxylic acid, cis-+/--3-boc-amino cyclohexanecarboxylic acid, boc-cis-3-aminocyclohexanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-rel-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino cyclohexanecarboxylic acid, cis-3-boc-amino cyclohexanecarboxylic acid, cis-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid
PubChem CID 16218560
IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 1 g
Formula Weight 243.30
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material cis-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid