missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-3-Nonen-1-ol 95.0+%, TCI America™
Supplier: TCI America N058810ML
Specifications
| cis-3-Nonen-1-ol | |
| Yellow | |
| C9H18O | |
| 10 mL | |
| IFTBJDZSLBRRMC-SREVYHEPSA-N | |
| (Z)-non-3-en-1-ol | |
| 5364631 | |
| ≥95.0% (GC) |
| 10340-23-5 | |
| 109°C | |
| MFCD00010314 | |
| cis-3-nonen-1-ol, z-non-3-en-1-ol, z-3-nonen-1-ol, 3-nonen-1-ol, z, unii-w8g7341qj8, z-3-nonenol, 3z-nonen-1-ol, 3z-3-nonen-1-ol, 3z-non-3-en-1-ol, 3 z ?-?nonenol | |
| CCCCCC=CCCO | |
| 142.242 | |
| 142.24 | |
| Liquid |
Chemical Identifiers
| 10340-23-5 | |
| 142.242 | |
| IFTBJDZSLBRRMC-SREVYHEPSA-N | |
| 5364631 | |
| CCCCCC=CCCO |
| C9H18O | |
| MFCD00010314 | |
| cis-3-nonen-1-ol, z-non-3-en-1-ol, z-3-nonen-1-ol, 3-nonen-1-ol, z, unii-w8g7341qj8, z-3-nonenol, 3z-nonen-1-ol, 3z-3-nonen-1-ol, 3z-non-3-en-1-ol, 3 z ?-?nonenol | |
| (Z)-non-3-en-1-ol |
Safety and Handling
EINECSNumber : (2)-0258
TSCA : Yes