missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2028806

 View more versions of this product

Catalog No. AAB2028806


Only null left
Explore available promotions

Description

Description

cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.

Solubility
Soluble in ethanol, ether. Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

cis-3-Methylcyclohexanol
73°C (163°F)
MFCD00067155
cis-3-methylcyclohexanol, cyclohexanol, 3-methyl-, cis, 1s,3r-3-methylcyclohexan-1-ol, 3-methyl cyclohexanol, cis, 3-methylcyclohexanol, z, cis 3-methylcyclohexanol, cis-3-methyl-cyclohexanol, 1s,3r-3-methyl-cyclohexanol, 1alpha-3alpha-methylcyclohexanol
HTSABYAWKQAHBT-RQJHMYQMSA-N
(1S,3R)-3-methylcyclohexan-1-ol
229503
95%
24965-90-0
C7H14O
5 g
Soluble in ethanol,ether. Slightly soluble in water.
CC1CCCC(C1)O
114.188
114.19

Chemical Identifiers

24965-90-0
114.188
HTSABYAWKQAHBT-RQJHMYQMSA-N
229503
CC1CCCC(C1)O
C7H14O
MFCD00067155
cis-3-methylcyclohexanol, cyclohexanol, 3-methyl-, cis, 1s,3r-3-methylcyclohexan-1-ol, 3-methyl cyclohexanol, cis, 3-methylcyclohexanol, z, cis 3-methylcyclohexanol, cis-3-methyl-cyclohexanol, 1s,3r-3-methyl-cyclohexanol, 1alpha-3alpha-methylcyclohexanol
(1S,3R)-3-methylcyclohexan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only