missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexen-1-yl 2-Methylbutyrate 97.0+%, TCI America™

Catalog No. M073425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M073425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M073425ML Supplier TCI America Supplier No. M073425ML
Only null left

Chemical Identifiers

CAS 53398-85-9
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00036530
InChI Key JKKGTSUICJWEKB-SREVYHEPSA-N
Synonym 2-Methylbutyric Acid cis-3-Hexen-1-yl Ester
PubChem CID 5365069
IUPAC Name [(Z)-hex-3-enyl] 2-methylbutanoate
SMILES CCC=CCCOC(=O)C(C)CC

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 184.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-yl 2-Methylbutyrate
EINECSNumber (2)-0760
TSCA Yes