missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexen-1-ol, 98%

Catalog No. AAA1031309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1031309 10 g
AAA1031318 50 g
AAA1031330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1031309 Supplier Thermo Scientific Chemicals Supplier No. A1031309

CAS: 928-96-1 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 928-96-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063217
InChI Key UFLHIIWVXFIJGU-ARJAWSKDSA-N
Synonym z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z
PubChem CID 5281167
ChEBI CHEBI:28857
SMILES CC\C=C/CCO

Specifications

Density 0.848
Boiling Point 156°C to 157°C
Flash Point 44°C (111°F)
Odor Pleasant
Refractive Index 1.44
Quantity 10 g
UN Number UN1987
Beilstein 1719712
Merck Index 14,4700
IUPAC Name (Z)-hex-3-en-1-ol
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material cis-3-Hexen-1-ol
Show More Show Less
Hazard Category H226-H319
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 213-192-8
RTECSNumber MP8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only