missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. p-7041485
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6228103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6228103 Supplier Thermo Scientific Chemicals Supplier No. H6228103

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 222530-33-8
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD01862295
InChI Key JSGHMGKJNZTKGF-BDAKNGLRSA-N
Synonym 1r,3s-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino-cyclohexanecarboxylic acid, cis-+/--3-boc-amino cyclohexanecarboxylic acid, boc-cis-3-aminocyclohexanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-rel-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3s-3-boc-amino cyclohexanecarboxylic acid, cis-3-boc-amino cyclohexanecarboxylic acid, cis-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 1r,3s-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid
PubChem CID 16218560
IUPAC Name (1R,3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Melting Point 143°C to 147°C
Quantity 1 g
Formula Weight 243.29
Percent Purity 97%
Chemical Name or Material cis-3-(Boc-amino)cyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only