missing translation for 'onlineSavingsMsg'
Learn More

cis-3-(Benzyloxymethyl)cyclobutanol, 97%, Thermo Scientific Chemicals

Catalog No. aah6257903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6257903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6257903 Supplier Thermo Scientific Chemicals Supplier No. H6257903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 172324-68-4 | Molecular Formula: C12H16O2 | Molecular Weight: 192.258 g/mol

cis-3-(Benzyloxymethyl)cyclobutanol is generally used as a intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-(Benzyloxymethyl)cyclobutanol is generally used as a intermediate in chemical research.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 172324-68-4
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD06657676
InChI Key KVQOEODTOJCIPA-UHFFFAOYSA-N
Synonym cis-3-benzyloxymethylcyclobutanol, trans-3-benzyloxymethylcyclobutanol, trans-3-benzyloxy methyl cyclobutanol, cis-3-benzyloxy methyl cyclobutanol, 1r,3r-3-benzyloxy methyl cyclobutanol, cyclobutanol,3-phenylmethoxy methyl-, cis, 3-benzyloxy methyl cyclobutan-1-ol, 1r,3r-3-benzyloxy methyl cyclobutan-1-ol
PubChem CID 10442570
IUPAC Name 3-(phenylmethoxymethyl)cyclobutan-1-ol
SMILES C1C(CC1O)COCC2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 192.25
Percent Purity 97%
Chemical Name or Material cis-3-(Benzyloxymethyl)cyclobutanol
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only