missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid, 95%

Catalog No. AC425900010
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425900010 1 g Glass bottle
AC425902500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425900010 Supplier Thermo Scientific Chemicals Supplier No. 425900010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22255-16-9 | C6H9NO2 | 127.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22255-16-9
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.14
MDL Number MFCD03093875,MFCD05663832
InChI Key JBDOTWVUXVXVDR-UOWFLXDJSA-N
Synonym 3-azabicyclo 3.1.0 hexane-2-carboxylicacid, 1s,2s,5r
PubChem CID 77906410
SMILES [O-]C(=O)[C@@H]1[NH2+]C[C@H]2C[C@@H]12

Specifications

Melting Point 252.0°C to 256.0°C
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
IUPAC Name (1R,2R,5S)-3-azabicyclo[3.1.0]hexan-3-ium-2-carboxylate
Formula Weight 127.14
Percent Purity 95%
Chemical Name or Material cis-3-Azabicyclo[3.1.0]hexane-2-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only