missing translation for 'onlineSavingsMsg'
Learn More

cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%

Catalog No. p-3777760
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC334270010 1 g Glass Bottle
AC334270050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC334270010 Supplier Thermo Scientific Chemicals Supplier No. 334270010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63216-49-9 | C12H20NO4 | 242.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63216-49-9
Molecular Formula C12H20NO4
Molecular Weight (g/mol) 242.30
MDL Number MFCD01863244,MFCD06410968
InChI Key QJEQJDJFJWWURK-BDAKNGLRSA-M
Synonym 1r,2s-boc-achc, boc-1,2-cis-achc-oh, boc-cis-2-aminocyclohexanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2s-boc-2-aminocyclohexane carboxylic acid, n-boc-+/--cis-2-amino-cyclohexane-carboxylic acid, cis-2-tert-butoxycarbonylamino-cyclohexanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, cis-1-t-butyloxycarbonylamino-cyclohexyl-2-carboxylic acid, 1r,2s-2-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid
PubChem CID 1512487
IUPAC Name (1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)N[C@H]1CCCC[C@H]1C([O-])=O

Specifications

Melting Point 127°C to 131°C
Color White
Flash Point 194°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula HOOCC6H10NHCOOC(CH3)3
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform
Formula Weight 243.3
Percent Purity 98%
Physical Form Amorphous Powder
Chemical Name or Material cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only