missing translation for 'onlineSavingsMsg'
Learn More

cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 136315-70-3
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
InChI Key BUEPEVBYNBQNED-SFYZADRCSA-N
Synonym 1r,2s-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-boc-amino cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-1-cyclopentanecarboxylic acid, cis-2-bocamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, +/--cis-2-boc-amino cyclopentanecarboxylic acid
PubChem CID 1268160
IUPAC Name (1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC1C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC299582500
View Documents
N/A 250mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

136315-70-3
229.28
1r,2s-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-boc-amino cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-1-cyclopentanecarboxylic acid, cis-2-bocamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, +/--cis-2-boc-amino cyclopentanecarboxylic acid
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
C11H19NO4
BUEPEVBYNBQNED-SFYZADRCSA-N
1268160
CC(C)(C)OC(=O)NC1CCCC1C(=O)O

Specifications

136315-70-3
98%
1r,2s-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-boc-amino cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonylamino-1-cyclopentanecarboxylic acid, cis-2-bocamino-cyclopentanecarboxylic acid, cis-2-tert-butoxycarbonyl amino cyclopentanecarboxylic acid, 1r,2s-2-tert-butoxycarbonyl amino cyclopentane-1-carboxylic acid, +/--cis-2-boc-amino cyclopentanecarboxylic acid
CC(C)(C)OC(=O)NC1CCCC1C(=O)O
229.28
229.28
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
Authentic
C11H19NO4
BUEPEVBYNBQNED-SFYZADRCSA-N
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopentane-1-carboxylic acid
1268160
98%
Videos
SDS
Documents

Documents

RUO – Research Use Only