missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Penten-1-ol 95.0+%, TCI America™

Catalog No. P119925ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P119925ML 25 mL
P11995ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P119925ML Supplier TCI America Supplier No. P119925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1576-95-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00063208
InChI Key BTSIZIIPFNVMHF-ARJAWSKDSA-N
Synonym cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol
PubChem CID 5364919
IUPAC Name (2Z)-pent-2-en-1-ol
SMILES CC\C=C/CO

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥95.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material cis-2-Penten-1-ol
EINECSNumber (2)-2393
TSCA Yes