Learn More
cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer
CAS: 1576-95-0 | C5H10O | 86.13 g/mol
Supplier: Thermo Scientific Chemicals A1426222
Description
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationscis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications
| cis-2-Penten-1-ol | |
| 0.853 | |
| 48°C (118°F) | |
| C5H10O | |
| MFCD00063208 | |
| UN1987 | |
| cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol | |
| BTSIZIIPFNVMHF-ARJAWSKDSA-N | |
| (Z)-pent-2-en-1-ol | |
| 5364919 | |
| 97% |
| 1576-95-0 | |
| 138°C | |
| remainder mainly trans-isomer | |
| 1.436 | |
| 100 g | |
| 1719473 | |
| Miscible with alcohol. Immiscible with water. | |
| CC\C=C/CO | |
| 86.13 | |
| 86.13 |
Chemical Identifiers
| 1576-95-0 | |
| 86.13 | |
| BTSIZIIPFNVMHF-ARJAWSKDSA-N | |
| 5364919 |
| C5H10O | |
| MFCD00063208 | |
| cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol | |
| CC\C=C/CO |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 216-415-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only