missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer

CAS: 1576-95-0 | C5H10O | 86.13 g/mol

Supplier:  Thermo Scientific Chemicals A1426214

 View more versions of this product

Catalog No. AAA1426214


Only null left
Explore available promotions

Description

Description

cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications

Specifications

cis-2-Penten-1-ol
0.853
48°C (118°F)
C5H10O
MFCD00063208
UN1987
cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol
BTSIZIIPFNVMHF-ARJAWSKDSA-N
(2Z)-pent-2-en-1-ol
5364919
97%
1576-95-0
138°C
remainder mainly trans-isomer
1.436
25 g
1719473
Miscible with alcohol. Immiscible with water.
CC\C=C/CO
86.13
86.13

Chemical Identifiers

1576-95-0
86.13
BTSIZIIPFNVMHF-ARJAWSKDSA-N
5364919
C5H10O
MFCD00063208
cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol
CC\C=C/CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

EINECSNumber : 216-415-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only