missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Penten-1-ol, 97%, remainder mainly trans-isomer

Catalog No. AAA1426206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1426206 5 g
AAA1426214 25 g
AAA1426222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1426206 Supplier Thermo Scientific Chemicals Supplier No. A1426206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1576-95-0 | C5H10O | 86.13 g/mol

cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-2-Penten-1-ol is used in the preparation of 1-bromo-pent-2-ene by reacting with phosphorus tribromide and pyridine. Further, it is a banana flavored compound.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 1576-95-0
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00063208
InChI Key BTSIZIIPFNVMHF-ARJAWSKDSA-N
Synonym cis-2-penten-1-ol, z-pent-2-en-1-ol, z-2-penten-1-ol, 2-penten-1-ol, z, e-2-penten-1-ol, unii-qnc2nb53mj, 2z-penten-1-ol, cis-2-pentenol, qnc2nb53mj, pent-2 e-enol
PubChem CID 5364919
SMILES CC\C=C/CO

Specifications

Density 0.853
Boiling Point 138°C
Flash Point 48°C (118°F)
Refractive Index 1.436
Quantity 5 g
UN Number UN1987
Beilstein 1719473
Solubility Information Miscible with alcohol. Immiscible with water.
IUPAC Name (2Z)-pent-2-en-1-ol
Formula Weight 86.13
Percent Purity 97%
Assay remainder mainly trans-isomer
Chemical Name or Material cis-2-Penten-1-ol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 216-415-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only