missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Butene-1,4-diol 94.0+%, TCI America™

Catalog No. B0693500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0693500G 500 g
B069325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0693500G Supplier TCI America Supplier No. B0693500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6117-80-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002924,MFCD00063207
InChI Key ORTVZLZNOYNASJ-UPHRSURJSA-N
Synonym cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
PubChem CID 643790
IUPAC Name (2Z)-but-2-ene-1,4-diol
SMILES OC\C=C/CO

Specifications

Melting Point 7°C
Color Yellow
Boiling Point 235°C
Quantity 500 g
Formula Weight 88.11
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2-Butene-1,4-diol
EINECSNumber (9)-1161
TSCA No