missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Aminocyclohexanol hydrochloride, 99+%, Thermo Scientific™

Catalog No. AC265850010
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC265850010 1g
AC265850050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC265850010 Supplier Thermo Scientific Chemicals Supplier No. 265850010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 6936-47-6
Molecular Formula C6H13NO·HCl
Molecular Weight (g/mol) 151.64
MDL Number MFCD00143981
InChI Key LKKCSUHCVGCGFA-RIHPBJNCSA-N
Synonym 1r,2s-2-aminocyclohexanol hydrochloride, cis-2-aminocyclohexanol hydrochloride, 1r,2s-2-aminocyclohexan-1-ol hydrochloride, 1r,2s-rel-2-aminocyclohexanol hydrochloride, cis 1r,2s-2-amino-cyclohexanol hydrochloride, 1r,2s-2-aminocyclohexanol hcl, cyclohexanol, 2-amino-, hydrochloride, cis, cis-2-aminocyclohexanolhydrochloride, cis-2-aminocyclohexanol hcl, cis-2-amino-cyclohexanol hydrochloride
PubChem CID 12228413
IUPAC Name (1R,2S)-2-aminocyclohexan-1-ol;hydrochloride
SMILES C1CCC(C(C1)N)O.Cl

Specifications

Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula H2NC6H10OH·HCl
Quantity 1g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,hot ethanol
Formula Weight 151.64
Percent Purity 99+%
Chemical Name or Material cis-2-Aminocyclohexanol hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only