missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylpiperazine 98.0+%, TCI America™

Catalog No. D236025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D236025G 25 g
D2360500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D236025G Supplier TCI America Supplier No. D236025G

Chemical Identifiers

CAS 21655-48-1
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00005956
InChI Key IFNWESYYDINUHV-OLQVQODUSA-N
Synonym cis-2,6-dimethylpiperazine, 2r,6s-2,6-dimethylpiperazine, cis-2,6-dimethyl piperazine, 2s,6r-2,6-dimethylpiperazine, cis-2,6-dimethyl-piperazine, piperazine, 2,6-dimethyl-, 2r,6s-rel, 2,6-dimethyl piperazine, 26dmprz, 2,6-dimethylpiperazine #, 2,6-cis-dimethylpiperazine
PubChem CID 6950261
IUPAC Name (2S,6R)-2,6-dimethylpiperazine
SMILES CC1CNCC(N1)C

Specifications

Melting Point 113°C
Color White-Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 1325
Formula Weight 114.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-2,6-Dimethylpiperazine