missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylmorpholine 97.0+%, TCI America™

Catalog No. p-7126409
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D074625ML 25 mL
D0746500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D074625ML Supplier TCI America Supplier No. D074625ML

Chemical Identifiers

CAS 6485-55-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00078428
InChI Key HNVIQLPOGUDBSU-OLQVQODUSA-N
Synonym cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624
PubChem CID 641500
IUPAC Name (2S,6R)-2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 mL
UN Number 1992
Formula Weight 115.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2,6-Dimethylmorpholine
RTECSNumber QE1750000
TSCA Yes