missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylmorpholine 97.0+%, TCI America™

Catalog No. d074625ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
D074625ML 25 mL
D0746500ML 500 mL
2 options

Catalog No. D074625ML

Supplier: TCI America D074625ML

Only null left

Chemical Identifiers

CAS 6485-55-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00078428
InChI Key HNVIQLPOGUDBSU-OLQVQODUSA-N
Synonym cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624
PubChem CID 641500
IUPAC Name (2S,6R)-2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 mL
UN Number 1992
Formula Weight 115.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-2,6-Dimethylmorpholine
RTECSNumber QE1750000
TSCA Yes