missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylmorpholine, 97%

Catalog No. AAL1300718
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1300718 50 g
AAL1300709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1300718 Supplier Thermo Scientific Chemicals Supplier No. L1300718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6485-55-8 | C6H13NO | 115.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6485-55-8
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00078428
InChI Key HNVIQLPOGUDBSU-OLQVQODUSA-N
Synonym cis-2,6-dimethylmorpholine, 2r,6s-2,6-dimethylmorpholine, 2s,6r-2,6-dimethylmorpholine, unii-vwd860p007, 2,6-dimethylmorpholine, cis, morpholine, 2,6-dimethyl-, cis, 2,6-dimethyl-morpholine, morpholine, 2,6-dimethyl-, 2r,6s-rel, cdmm, pubchem16624
PubChem CID 641500
IUPAC Name (2S,6R)-2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Melting Point -85°C
Density 0.935
Boiling Point 146°C to 147°C
Flash Point 48°C (118°F)
Refractive Index 1.446
Quantity 50 g
UN Number UN1992
Beilstein 878182
Formula Weight 115.18
Percent Purity 97%
Chemical Name or Material cis-2,6-Dimethylmorpholine
Show More Show Less
Hazard Category H226-H302-H311-H314-H335
Hazard Statement GHS H Statement
H318-H226-H312
Causes serious eye damage.
Flammable liquid and vapor.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 229-353-0
RTECSNumber QE1750000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only