missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-13-Docosenol 98.0+%, TCI America™
$55.92 - $158.44
Chemical Identifiers
| CAS | 629-98-1 |
|---|---|
| Molecular Formula | C22H44O |
| Molecular Weight (g/mol) | 324.59 |
| MDL Number | MFCD00055936 |
| InChI Key | CFOQKXQWGLAKSK-KTKRTIGZSA-N |
| Synonym | Erucyl Alcohol |
| PubChem CID | 5354168 |
| ChEBI | CHEBI:77416 |
| IUPAC Name | (13Z)-docos-13-en-1-ol |
| SMILES | CCCCCCCC\C=C/CCCCCCCCCCCCO |
Chemical Identifiers
| 629-98-1 | |
| 324.59 | |
| CFOQKXQWGLAKSK-KTKRTIGZSA-N | |
| 5354168 | |
| (13Z)-docos-13-en-1-ol |
| C22H44O | |
| MFCD00055936 | |
| Erucyl Alcohol | |
| CHEBI:77416 | |
| CCCCCCCC\C=C/CCCCCCCCCCCCO |
Specifications
| 629-98-1 | |
| White-Yellow | |
| C22H44O | |
| 1 g | |
| CFOQKXQWGLAKSK-KTKRTIGZSA-N | |
| (13Z)-docos-13-en-1-ol | |
| 5354168 | |
| 324.59 | |
| Crystals |
| 33°C | |
| 225°C | |
| MFCD00055936 | |
| Erucyl Alcohol | |
| CCCCCCCC\C=C/CCCCCCCCCCCCO | |
| 324.59 | |
| CHEBI:77416 | |
| ≥98.0% (GC) | |
| cis-13-Docosenol |
Safety and Handling
TSCA : No