missing translation for 'onlineSavingsMsg'
Learn More

cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene, 94%

Catalog No. AAH5125806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5125806 5 g
AAH5125803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH5125806 Supplier Thermo Scientific Chemicals Supplier No. H5125806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 64724-29-4 | C16H34OSn | 361.157 g/mol

cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.

Solubility
Soluble in water.

Notes
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is to be stored in a cool, dry place and is to be sealed properly.

Chemical Identifiers

CAS 64724-29-4
Molecular Formula C16H34OSn
Molecular Weight (g/mol) 361.157
MDL Number MFCD07787391
InChI Key WARKYKQCOXTIAO-UHFFFAOYSA-N
Synonym z-1-ethoxy-2-tributylstannyl ethene, tributyl z-2-ethoxyvinyl stannane, tributyl z-2-ethoxyethenyl stannane, tributyl-z-2-ethoxyethenyl stannane, z-tributyl 2-ethoxyvinyl stannane, cis-1-ethoxy-2-tri-n-butylstannyl ethylene, cis-tributyl 2-ethoxyethenyl stannane, tributyl-z-2-ethoxyvinyl stannane, z-1-ethoxy-2-tributylstannyl ethylene
PubChem CID 5706773
IUPAC Name tributyl-[(Z)-2-ethoxyethenyl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=COCC

Specifications

Melting Point 84°C to 87°C
Density 1.082
Quantity 5 g
UN Number UN2788
Solubility Information Soluble in water.
Formula Weight 361.15
Percent Purity 94%
Chemical Name or Material cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only