missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane, 97%, Thermo Scientific Chemicals
$1,246.52
Chemical Identifiers
| CAS | 1069120-19-9 |
|---|---|
| Molecular Formula | C13H25NO3 |
| Molecular Weight (g/mol) | 243.35 |
| MDL Number | MFCD03844626,MFCD03844625 |
| InChI Key | KTNFSGIXLVLQNK-UHFFFAOYSA-N |
| Synonym | tert-butyl trans-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl n-1r,4r-4-2-hydroxyethyl cyclohexyl carbamate, ktnfsgixlvlqnk-xypyzodxsa-n, butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl n-4-2-hydroxyethyl cyclohexyl carbamate |
| PubChem CID | 24720929 |
| IUPAC Name | tert-butyl N-[4-(2-hydroxyethyl)cyclohexyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCC(CCO)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH6255703
|
N/A | 1 g |
Each for $1,246.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1069120-19-9 | |
| 243.35 | |
| KTNFSGIXLVLQNK-UHFFFAOYSA-N | |
| 24720929 | |
| CC(C)(C)OC(=O)NC1CCC(CCO)CC1 |
| C13H25NO3 | |
| MFCD03844626,MFCD03844625 | |
| tert-butyl trans-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl n-1r,4r-4-2-hydroxyethyl cyclohexyl carbamate, ktnfsgixlvlqnk-xypyzodxsa-n, butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl n-4-2-hydroxyethyl cyclohexyl carbamate | |
| tert-butyl N-[4-(2-hydroxyethyl)cyclohexyl]carbamate |
Specifications
| 1069120-19-9 | |
| MFCD03844626,MFCD03844625 | |
| KTNFSGIXLVLQNK-UHFFFAOYSA-N | |
| tert-butyl N-[4-(2-hydroxyethyl)cyclohexyl]carbamate | |
| 24720929 | |
| 97% |
| C13H25NO3 | |
| tert-butyl trans-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl-cyclohexylcarbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl trans-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl cis-4-2-hydroxyethyl cyclohexylcarbamate, tert-butyl n-1r,4r-4-2-hydroxyethyl cyclohexyl carbamate, ktnfsgixlvlqnk-xypyzodxsa-n, butyl trans-4-2-hydroxyethyl cyclohexyl carbamate, tert-butyl n-4-2-hydroxyethyl cyclohexyl carbamate | |
| CC(C)(C)OC(=O)NC1CCC(CCO)CC1 | |
| 243.35 | |
| 243.34 | |
| cis-1-(Boc-amino)-4-(2-hydroxyethyl)cyclohexane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only