missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,4-Dimethylcyclohexane 98.0+%, TCI America™
Supplier: TCI America D171525ML
Specifications
| cis-1,4-Dimethylcyclohexane | |
| Colorless | |
| C8H16 | |
| 25 mL | |
| QRMPKOFEUHIBNM-UHFFFAOYSA-N | |
| 1,4-dimethylcyclohexane | |
| 11523 | |
| ≥98.0% (GC) |
| 624-29-3 | |
| 125°C | |
| MFCD00001508,MFCD00064174,MFCD00064955 | |
| 2263 | |
| CC1CCC(C)CC1 | |
| 112.22 | |
| 112.22 | |
| Liquid |
Chemical Identifiers
| 624-29-3 | |
| 112.22 | |
| QRMPKOFEUHIBNM-UHFFFAOYSA-N | |
| 1,4-dimethylcyclohexane |
| C8H16 | |
| MFCD00001508,MFCD00064174,MFCD00064955 | |
| 11523 | |
| CC1CCC(C)CC1 |
Safety and Handling
EINECSNumber : (3)-2227
RTECSNumber : GV0200000
TSCA : No