missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Dimethylcyclohexane 98.0+%, TCI America™

Catalog No. D17155ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17155ML 5 mL
D171525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17155ML Supplier TCI America Supplier No. D17155ML
Only null left

Chemical Identifiers

CAS 624-29-3
Molecular Formula C8H16
Molecular Weight (g/mol) 112.22
MDL Number MFCD00001508,MFCD00064174,MFCD00064955
InChI Key QRMPKOFEUHIBNM-UHFFFAOYSA-N
PubChem CID 11523
IUPAC Name 1,4-dimethylcyclohexane
SMILES CC1CCC(C)CC1

Specifications

Color Colorless
Boiling Point 125°C
Quantity 5 mL
UN Number 2263
Formula Weight 112.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-1,4-Dimethylcyclohexane
EINECSNumber (3)-2227
RTECSNumber GV0200000
TSCA No