missing translation for 'onlineSavingsMsg'
Learn More

cis-1,3-Pentadiene (stabilized with TBC) 97.0+%, TCI America™

Catalog No. p06505g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06505G Supplier TCI America Supplier No. P06505G
Only null left

Specifications

CAS 1574-41-0
Melting Point -140°C
Color Colorless
Boiling Point 43°C
Molecular Formula C5H8
Quantity 5 g
MDL Number MFCD00063977
UN Number 3295
Synonym cis-Piperylene
InChI Key PMJHHCWVYXUKFD-PLNGDYQASA-N
SMILES C\C=C/C=C
IUPAC Name (3Z)-penta-1,3-diene
Molecular Weight (g/mol) 68.12
PubChem CID 643785
Formula Weight 68.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-1,3-Pentadiene (stabilized with TBC)
Show More Show Less
EINECSNumber (2)-0020
RTECSNumber RZ2469000
TSCA Yes
Recommended Storage Freezer