missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,3-Dimethylcyclohexane 99.0+%, TCI America™
$154.71 - $410.10
Chemical Identifiers
| CAS | 638-04-0 |
|---|---|
| Molecular Formula | C8H16 |
| Molecular Weight (g/mol) | 112.22 |
| MDL Number | MFCD00001507 |
| InChI Key | SGVUHPSBDNVHKL-OCAPTIKFSA-N |
| Synonym | cis-1,3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexane, cyclohexane, 1,3-dimethyl-, cis, 1,cis-3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexan, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, cis-hexahydro-m-xylene, c-1,3-dimethylcyclohexane, 1,3-dimethyl cis-cyclohexane |
| PubChem CID | 252361 |
| IUPAC Name | (1R,3S)-1,3-dimethylcyclohexane |
| SMILES | C[C@H]1CCC[C@@H](C)C1 |
Chemical Identifiers
| 638-04-0 | |
| 112.22 | |
| SGVUHPSBDNVHKL-OCAPTIKFSA-N | |
| 252361 | |
| C[C@H]1CCC[C@@H](C)C1 |
| C8H16 | |
| MFCD00001507 | |
| cis-1,3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexane, cyclohexane, 1,3-dimethyl-, cis, 1,cis-3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexan, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, cis-hexahydro-m-xylene, c-1,3-dimethylcyclohexane, 1,3-dimethyl cis-cyclohexane | |
| (1R,3S)-1,3-dimethylcyclohexane |
Specifications
| 638-04-0 | |
| 120°C | |
| MFCD00001507 | |
| 2263 | |
| SGVUHPSBDNVHKL-OCAPTIKFSA-N | |
| (1R,3S)-1,3-dimethylcyclohexane | |
| 252361 | |
| ≥99.0% (GC) | |
| cis-1,3-Dimethylcyclohexane |
| Colorless | |
| C8H16 | |
| 5 mL | |
| cis-1,3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexane, cyclohexane, 1,3-dimethyl-, cis, 1,cis-3-dimethylcyclohexane, 1r,3s-1,3-dimethylcyclohexan, unii-9sgo12qv70, 1,3-dimethylcyclohexane, cis, cis-hexahydro-m-xylene, c-1,3-dimethylcyclohexane, 1,3-dimethyl cis-cyclohexane | |
| C[C@H]1CCC[C@@H](C)C1 | |
| 112.22 | |
| 112.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2227
TSCA : No